1-Butyl-3-methylimidazoliumbromid 97%

Synonyme: 3-Butyl-1-methyl-1H-imidazol-3-ium bromide

BD159375-100G BD159375-25G BD159375-5G BD159375-10G BD159375-1000G BD159375-500G
BLDPBD159375-100GEA 40 EUR
BLDPBD159375-100G BLDPBD159375-25G BLDPBD159375-5G BLDPBD159375-10G BLDPBD159375-1000G BLDPBD159375-500G
1-Butyl-3-methylimidazoliumbromid 97%
1-Butyl-3-methylimidazolium bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₅BrN₂
Molecular Weight: 219,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD03427611
CAS-Nummer: 85100-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR