(6-Methoxypyridin-3-yl)methylamine 98%

BLDPBD159371-1GEA 73 EUR
BLDPBD159371-1G BLDPBD159371-25G BLDPBD159371-10G BLDPBD159371-250MG BLDPBD159371-5G
(6-Methoxypyridin-3-yl)methylamine 98%
(6-Methoxypyridin-3-yl)methylamine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₁₀N₂O
Molecular Weight: 138,17 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD06212825
CAS-Nummer: 262295-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR