2-Bromo-5-fluorophenylacetonitrile 98%

Synonyme: 2-Bromo-5-fluorobenzyl cyanide,2-(2-Bromo-5-fluorophenyl)acetonitrile,(2-Bromo-5-fluorophenyl)acetonitrile

BD159346-250MG BD159346-25G BD159346-1G BD159346-100G BD159346-5G BD159346-10G
BLDPBD159346-250MGEA 9 EUR
BLDPBD159346-250MG BLDPBD159346-25G BLDPBD159346-1G BLDPBD159346-100G BLDPBD159346-5G BLDPBD159346-10G
2-Bromo-5-fluorophenylacetonitrile 98%
2-Bromo-5-fluorophenylacetonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅BrFN
Molecular Weight: 214,034 g/mol
Lagertemperatur: Ambient
MDL: MFCD03094439
CAS-Nummer: 886761-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR