4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile 97%

BLDPBD159307-1GEA 415 EUR
BLDPBD159307-1G BLDPBD159307-100MG BLDPBD159307-250MG
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile 97%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₄ClN₃
Lagertemperatur: Kühlschrank
MDL: MFCD09965877
CAS-Nummer: 920966-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR