5-Chloropyrazine-2-carbonitrile 97%

BLDPBD159299-1GEA 54 EUR
BLDPBD159299-1G BLDPBD159299-10G BLDPBD159299-5G BLDPBD159299-25G BLDPBD159299-100G BLDPBD159299-250MG
5-Chloropyrazine-2-carbonitrile 97%
5-Chloropyrazine-2-carbonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C5H2ClN3
Molecular Weight: 139,5425 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09607700
CAS-Nummer: 36070-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR