1-Brom-4-(difluormethyl)benzol 95%

Synonyme: 4-Bromobenzal fluoride

BD159297-1G BD159297-25G BD159297-250MG BD159297-5G
BLDPBD159297-1GEA 15 EUR
BLDPBD159297-1G BLDPBD159297-25G BLDPBD159297-250MG BLDPBD159297-5G
1-Brom-4-(difluormethyl)benzol 95%
1-Bromo-4-(difluoromethyl)benzene

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BrF₂
Molecular Weight: 207,02 g/mol
Lagertemperatur: Ambient
MDL: MFCD06657964
CAS-Nummer: 51776-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR