5-Brom-m-toluylsäure 98%

Synonyme: 3-Bromo-5-methylbenzoic acid

BD159260-5G BD159260-25G BD159260-500G BD159260-10G BD159260-100G BD159260-1G
BLDPBD159260-5GEA 25 EUR
BLDPBD159260-5G BLDPBD159260-25G BLDPBD159260-500G BLDPBD159260-10G BLDPBD159260-100G BLDPBD159260-1G
5-Brom-m-toluylsäure 98%
5-Brom-m-toluylsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrO₂
Molecular Weight: 215,0440 g/mol
Lagertemperatur: Ambient
MDL: MFCD02684926
CAS-Nummer: 58530-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR