4-Amino-2-chloro-3-methylbenzonitrile 97%

BLDPBD159253-1GEA 348 EUR
BLDPBD159253-1G BLDPBD159253-100MG BLDPBD159253-250MG
4-Amino-2-chloro-3-methylbenzonitrile 97%
4-Amino-2-chloro-3-methylbenzonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₇ClN₂
Molecular Weight: 166,60 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08059126
CAS-Nummer: 573768-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR