6-Fluoropyridine-2-boronic acid 95%

Synonyme: (6-Fluoropyridin-2-yl)boronic acid

BD159243-250MG BD159243-1G
BLDPBD159243-250MGEA 112 EUR
BLDPBD159243-250MG BLDPBD159243-1G
6-Fluoropyridine-2-boronic acid 95%
6-Fluoropyridine-2-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₅H₅BFNO₂
Molecular Weight: 140,908 g/mol
Lagertemperatur: Freezer
MDL: MFCD08705713
CAS-Nummer: 916176-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR