6-Bromoimidazo[1,2-a]pyrazin-8-amine 97%

Synonyme: 8-Amino-6-bromoimidazo[1,2-a]pyrazine

BD159213-100MG BD159213-5G BD159213-1G BD159213-10G BD159213-250MG
BLDPBD159213-100MGEA 18 EUR
BLDPBD159213-100MG BLDPBD159213-5G BLDPBD159213-1G BLDPBD159213-10G BLDPBD159213-250MG
6-Bromoimidazo[1,2-a]pyrazin-8-amine 97%
6-Bromoimidazo[1,2-a]pyrazin-8-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₅BrN₄
Molecular Weight: 213,0400 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00874115
CAS-Nummer: 117718-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR