5-Fluorobenzo[d]isoxazol-3-amine 95%

BD159161-250MG BD159161-1G
BLDPBD159161-250MGEA 95 EUR
BLDPBD159161-250MG BLDPBD159161-1G
5-Fluorobenzo[d]isoxazol-3-amine 95%
5-Fluorobenzo[d]isoxazol-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C7H5FN2O
Molecular Weight: 152,1300 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09999137
CAS-Nummer: 868271-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR