tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate 95%

Synonyme: N-t-BOC-4-Methylene-3-Piperidinol,tert-Butyl 3-hydroxy-4-methylenepiperidine-1-carboxylate

BD159151-250MG BD159151-1G BD159151-100MG BD159151-5G
BLDPBD159151-250MGEA 66 EUR
BLDPBD159151-250MG BLDPBD159151-1G BLDPBD159151-100MG BLDPBD159151-5G
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate 95%
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,2735 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD15474939
CAS-Nummer: 159635-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR