5-Bromo-2-ethylpyridine 98%

BD159133-1G BD159133-100MG BD159133-250MG BD159133-5G
BLDPBD159133-1GEA 137 EUR
BLDPBD159133-1G BLDPBD159133-100MG BLDPBD159133-250MG BLDPBD159133-5G
5-Bromo-2-ethylpyridine 98%
5-Bromo-2-ethylpyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₈BrN
Molecular Weight: 186,0491 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11226235
CAS-Nummer: 38749-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR