5,6,7,8-Tetrahydro-2,6-naphthyridin-1(2H)-one 95%

BD159107-100MG
BLDPBD159107-100MGEA 200 EUR
BLDPBD159107-100MG
5,6,7,8-Tetrahydro-2,6-naphthyridin-1(2H)-one 95%
5,6,7,8-Tetrahydro-2,6-naphthyridin-1(2H)-one

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09999259
CAS-Nummer: 1170830-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR