4-Iodisoxazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD159090-250MGEA 24 EUR
BLDPBD159090-250MG BLDPBD159090-1G BLDPBD159090-5G
4-Iodisoxazol 98%
4-Iodoisoxazole

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formel: C₃H₂INO
Molecular Weight: 194,9600 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09999222
CAS-Nummer: 847490-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR