4-Chloroquinoline-7-carboxylic acid 98%

BD159076-100MG BD159076-250MG BD159076-5G BD159076-1G
BLDPBD159076-100MGEA 40 EUR
BLDPBD159076-100MG BLDPBD159076-250MG BLDPBD159076-5G BLDPBD159076-1G
4-Chloroquinoline-7-carboxylic acid 98%
4-Chloroquinoline-7-carboxylic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,61 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11518966
CAS-Nummer: 49713-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR