3-Amino-4-fluor-1-methyl-1H-indazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD159023-1GEA 176 EUR
BLDPBD159023-1G BLDPBD159023-250MG BLDPBD159023-5G BLDPBD159023-100MG
3-Amino-4-fluor-1-methyl-1H-indazol 98%
3-Amino-4-fluoro-1-methyl-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₈FN₃
Molecular Weight: 165,1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11109397
CAS-Nummer: 162502-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR