4-Glycidyloxycarbazole 98%

Synonyme: 4-(2,3-Epoxypropoxy)carbazole

BD15900-5G
BLDPBD15900-5GEA 8 EUR
BLDPBD15900-5G
4-Glycidyloxycarbazole 98%
4-Glycidyloxycarbazole
Formel: C₁₅H₁₃NO₂
Molecular Weight: 239,27 g/mol
MDL: MFCD03411884
CAS-Nummer: 51997-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR