Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%

Synonyme: Ethyl 4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

BD158984-100MG BD158984-1G BD158984-250MG BD158984-50MG
BLDPBD158984-100MGEA 121 EUR
BLDPBD158984-100MG BLDPBD158984-1G BLDPBD158984-250MG BLDPBD158984-50MG
Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%
Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₉N₃O₃
Molecular Weight: 207,19 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10699251
CAS-Nummer: 187724-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR