Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate 97%

Synonyme: Ethyl 4-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate

BD158977-100MG BD158977-5G BD158977-1G BD158977-250MG
BLDPBD158977-100MGEA 24 EUR
BLDPBD158977-100MG BLDPBD158977-5G BLDPBD158977-1G BLDPBD158977-250MG
Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate 97%
Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈ClN₃O₂
Molecular Weight: 225,6317 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11518913
CAS-Nummer: 144927-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR