7-Benzyloxyindol 98%

Synonyme: 7-(Benzyloxy)-1H-indole

BD15895-1G BD15895-5G BD15895-10G BD15895-25G
BLDPBD15895-1GEA 18 EUR
BLDPBD15895-1G BLDPBD15895-5G BLDPBD15895-10G BLDPBD15895-25G
7-Benzyloxyindol 98%
7-Benzyloxyindol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
Lagertemperatur: Ambient
MDL: MFCD00037974
CAS-Nummer: 20289-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR