7-tert-Butyl-3-ethyl-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-3,7(8H)-dicarboxylate 95%

BLDPBD158930-250MGEA 51 EUR
BLDPBD158930-250MG BLDPBD158930-1G BLDPBD158930-100MG
7-tert-Butyl-3-ethyl-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-3,7(8H)-dicarboxylate 95%
7-tert-Butyl-3-ethyl-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-3,7(8H)-dicarboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₂₀N₄O₄
Molecular Weight: 296,3200 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11616512
CAS-Nummer: 1215852-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR