7-Iodo-1H-indazole 97%

BD158924-100MG BD158924-250MG BD158924-5G BD158924-1G
BLDPBD158924-100MGEA 33 EUR
BLDPBD158924-100MG BLDPBD158924-250MG BLDPBD158924-5G BLDPBD158924-1G
7-Iodo-1H-indazole 97%
7-Iodo-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₅IN₂
Molecular Weight: 244,03 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09263215
CAS-Nummer: 945761-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR