Ethylthiazol-2-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD158840-1GEA 42 EUR
BLDPBD158840-1G BLDPBD158840-100MG BLDPBD158840-250MG BLDPBD158840-5G
Ethylthiazol-2-carboxylat 97%
Ethylthiazol-2-carboxylat

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₇NO₂S
Molecular Weight: 157,1900 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06204689
CAS-Nummer: 14527-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR