4-Chloropyrrolo[1,2-f][1,2,4]triazine 95%

BLDPBD158796-1GEA 360 EUR
BLDPBD158796-1G BLDPBD158796-5G BLDPBD158796-250MG BLDPBD158796-100MG
4-Chloropyrrolo[1,2-f][1,2,4]triazine 95%
4-Chloropyrrolo[1,2-f][1,2,4]triazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄ClN₃
Molecular Weight: 153,57 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11519387
CAS-Nummer: 888720-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR