4-Cyan-2-iodanilin 98%

Synonyme: 4-Amino-3-iodbenzoesäurenitril,4-Amino-3-iodbenzonitril

BD158777-5G BD158777-25G BD158777-1G BD158777-10G BD158777-500G BD158777-100G
BLDPBD158777-5GEA 22 EUR
BLDPBD158777-5G BLDPBD158777-25G BLDPBD158777-1G BLDPBD158777-10G BLDPBD158777-500G BLDPBD158777-100G
4-Cyan-2-iodanilin 98%
4-Cyan-2-iodanilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅IN₂
Molecular Weight: 244,03 g/mol
Lagertemperatur: Ambient
MDL: MFCD04039964
CAS-Nummer: 33348-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR