4-Methoxypyrimidine-2-carboxylic acid 98%

BD158771-1G BD158771-100MG BD158771-250MG
BLDPBD158771-1GEA 681 EUR
BLDPBD158771-1G BLDPBD158771-100MG BLDPBD158771-250MG
4-Methoxypyrimidine-2-carboxylic acid 98%
4-Methoxypyrimidine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₆N₂O₃
Molecular Weight: 154,12 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11520492
CAS-Nummer: 1208682-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR