(R)-2-Methoxypropan-1-ol 95%

BD158755-100MG BD158755-250MG
BLDPBD158755-100MGEA 92 EUR
BLDPBD158755-100MG BLDPBD158755-250MG
(R)-2-Methoxypropan-1-ol 95%
(R)-2-Methoxypropan-1-ol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
MDL: MFCD01632586
CAS-Nummer: 6131-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR