Ethyl-2-formyl-3-oxopropionate 97%

Synonyme: Ethyl 2-formyl-3-oxopropanoate,Ethyl 2-Formyl-3-oxopropionate,Diformylacetic acid ethyl ester,2-Formyl-3-oxopropionic acid ethyl ester,Ethyl diformylacetate

BD158749-100G BD158749-25G BD158749-1G BD158749-5G BD158749-10G
BLDPBD158749-100GEA 331 EUR
BLDPBD158749-100G BLDPBD158749-25G BLDPBD158749-1G BLDPBD158749-5G BLDPBD158749-10G
Ethyl-2-formyl-3-oxopropionate 97%
Ethyl-2-formyl-3-oxopropionate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₆H₈O₄
Molecular Weight: 144,13 g/mol
Lagertemperatur: Freezer
MDL: MFCD11112084
CAS-Nummer: 80370-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR