2-Aminothiophene-3-carboxylic acid 97%

BLDPBD158747-100MGEA 72 EUR
BLDPBD158747-100MG
2-Aminothiophene-3-carboxylic acid 97%
2-Aminothiophene-3-carboxylic acid
Formel: C₅H₅NO₂S
MDL: MFCD03426102
CAS-Nummer: 56387-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR