5-Chloropyridine-3,4-diamine 97%

Synonyme: 5-Chloro-3,4-diaminopyridine

BD158712-5G BD158712-1G BD158712-25G BD158712-250MG
BLDPBD158712-5GEA 292 EUR
BLDPBD158712-5G BLDPBD158712-1G BLDPBD158712-25G BLDPBD158712-250MG
5-Chloropyridine-3,4-diamine 97%
5-Chloropyridine-3,4-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C5H6ClN3
Molecular Weight: 143,57 g/mol
Lagertemperatur: Ambient
MDL: MFCD09263519
CAS-Nummer: 929074-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR