3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid 98%

BD158683-1G BD158683-5G BD158683-250MG BD158683-100MG
BLDPBD158683-1GEA 159 EUR
BLDPBD158683-1G BLDPBD158683-5G BLDPBD158683-250MG BLDPBD158683-100MG
3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid 98%
3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₅H₂₅N₇O₃
Molecular Weight: 471,51 g/mol
Lagertemperatur: Ambient
MDL: MFCD09837830
CAS-Nummer: 211914-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR