1-Chlor-5-nitroisochinolin 97%

BD158670-10G BD158670-5G BD158670-250MG BD158670-25G BD158670-1G BD158670-100MG
BLDPBD158670-10GEA 430 EUR
BLDPBD158670-10G BLDPBD158670-5G BLDPBD158670-250MG BLDPBD158670-25G BLDPBD158670-1G BLDPBD158670-100MG
1-Chlor-5-nitroisochinolin 97%
1-Chloro-5-nitroisoquinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₅ClN₂O₂
Molecular Weight: 208,6000 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11052590
CAS-Nummer: 58142-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR