5,6-Dichlor-1H-indol 98%

BLDPBD158633-5GEA 46 EUR
BLDPBD158633-5G BLDPBD158633-10G BLDPBD158633-100G BLDPBD158633-1G BLDPBD158633-25G
5,6-Dichlor-1H-indol 98%
5,6-Dichloro-1H-indole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₅Cl₂N
Molecular Weight: 186,0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075036
CAS-Nummer: 121859-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR