1-(2-Bromophenyl)ethylamine 97%

BD158629-250MG BD158629-1G BD158629-100MG BD158629-5G
BLDPBD158629-250MGEA 66 EUR
BLDPBD158629-250MG BLDPBD158629-1G BLDPBD158629-100MG BLDPBD158629-5G
1-(2-Bromophenyl)ethylamine 97%
1-(2-Bromophenyl)ethylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD05215385
CAS-Nummer: 113899-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR