4-Azaindole-3-carboxylic acid 97%

BLDPBD158625-1GEA 70 EUR
BLDPBD158625-1G BLDPBD158625-100MG BLDPBD158625-5G BLDPBD158625-250MG
4-Azaindole-3-carboxylic acid 97%
4-Azaindole-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₆N₂O₂
Molecular Weight: 162,1454 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11559014
CAS-Nummer: 860496-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR