4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine 95%

BD158601-1G BD158601-250MG BD158601-100MG
BLDPBD158601-1GEA 1365 EUR
BLDPBD158601-1G BLDPBD158601-250MG BLDPBD158601-100MG
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
MDL: MFCD09880145
CAS-Nummer: 1000340-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR