4-Chlor-3-(dimethylcarbamoyl)phenylboronsäure 98%

BLDPBD158598-5GEA 486 EUR
BLDPBD158598-5G BLDPBD158598-250MG BLDPBD158598-1G
4-Chlor-3-(dimethylcarbamoyl)phenylboronsäure 98%
4-Chlor-3-(dimethylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₁BClNO₃
Molecular Weight: 227,4500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07783868
CAS-Nummer: 871332-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR