3-Fluor-5-(methylcarbamoyl)phenylboronsäure 98%

Synonyme: (3-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid,3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

BD158585-1G BD158585-5G BD158585-250MG
BLDPBD158585-1GEA 138 EUR
BLDPBD158585-1G BLDPBD158585-5G BLDPBD158585-250MG
3-Fluor-5-(methylcarbamoyl)phenylboronsäure 98%
3-Fluor-5-(methylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₉BFNO₃
Molecular Weight: 196,97 g/mol
Lagertemperatur: Ambient
MDL: MFCD07363793
CAS-Nummer: 871332-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR