(S)-4-Hydroxypiperidin-2-one 95%

BD158580-100MG BD158580-250MG
BLDPBD158580-100MGEA 110 EUR
BLDPBD158580-100MG BLDPBD158580-250MG
(S)-4-Hydroxypiperidin-2-one 95%
(S)-4-Hydroxypiperidin-2-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD09752973
CAS-Nummer: 476014-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR