5-Iodopyrimidin-4-ol 98%

Synonyme: 4-Hydroxy-5-iodopyrimidine

BD158507-250MG BD158507-100MG BD158507-5G BD158507-1G
BLDPBD158507-250MGEA 26 EUR
BLDPBD158507-250MG BLDPBD158507-100MG BLDPBD158507-5G BLDPBD158507-1G
5-Iodopyrimidin-4-ol 98%
5-Iodopyrimidin-4-ol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C4H3IN2O
Molecular Weight: 221,98 g/mol
Lagertemperatur: Ambient
CAS-Nummer: 4349-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR