4-Bromo-2-(trifluoromethoxy)benzoic acid 95%

BLDPBD158497-10GEA 51 EUR
BLDPBD158497-10G BLDPBD158497-5G BLDPBD158497-25G BLDPBD158497-1G
4-Bromo-2-(trifluoromethoxy)benzoic acid 95%
4-Bromo-2-(trifluoromethoxy)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₄BrF₃O₃
Molecular Weight: 285,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546979
CAS-Nummer: 509142-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR