Pyrido[3,4-d]pyrimidin-4(3H)-one 95%

BD158432-250MG BD158432-100MG BD158432-1G
BLDPBD158432-250MGEA 72 EUR
BLDPBD158432-250MG BLDPBD158432-100MG BLDPBD158432-1G
Pyrido[3,4-d]pyrimidin-4(3H)-one 95%
Pyrido[3,4-d]pyrimidin-4(3H)-one

Storage condition: Sealed in dry, Room Temperature

Formel: C7H5N3O
Molecular Weight: 147,1300 g/mol
Lagertemperatur: Ambient
MDL: MFCD09999183
CAS-Nummer: 19178-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR