Methyl-5-methyl-4-oxazolcarboxylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD158406-1GEA 88 EUR
BLDPBD158406-1G BLDPBD158406-250MG
Methyl-5-methyl-4-oxazolcarboxylat 98%
Methyl-5-methyl-4-oxazolcarboxylat

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₇NO₃
Molecular Weight: 141,1247 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08689502
CAS-Nummer: 41172-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR