2-Chlorothiophene-3-carboxylic acid 98%

BD158350-100MG BD158350-5G BD158350-1G BD158350-250MG
BLDPBD158350-100MGEA 10 EUR
BLDPBD158350-100MG BLDPBD158350-5G BLDPBD158350-1G BLDPBD158350-250MG
2-Chlorothiophene-3-carboxylic acid 98%
2-Chlorothiophene-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₃ClO₂S
Molecular Weight: 162,5941 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00098423
CAS-Nummer: 53935-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR