tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate 95%

BD158307-250MG BD158307-100MG
BLDPBD158307-250MGEA 13 EUR
BLDPBD158307-250MG BLDPBD158307-100MG
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate 95%
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₃BrN₂O₂S
Molecular Weight: 293,1800 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09878588
CAS-Nummer: 1000576-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR