5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%

Synonyme: tert-Butyl 2-(methylsulphonyl)-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate,tert-Butyl 5,7-dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine-6-carboxylate

BD158305-1G BD158305-100MG BD158305-5G BD158305-250MG
BLDPBD158305-1GEA 423 EUR
BLDPBD158305-1G BLDPBD158305-100MG BLDPBD158305-5G BLDPBD158305-250MG
5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%
5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₇N₃O₄S
Molecular Weight: 299,346 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09839781
CAS-Nummer: 365996-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR