tert-Butyl-2,4-dioxopiperidine-1-carboxylate 97%

Synonyme: tert-Butyl 2,4-dioxopiperidine-1-carboxylate,Piperidine-2,4-dione, N-BOC protected

BD158304-5G BD158304-100G BD158304-1G BD158304-25G BD158304-10G BD158304-500G
BLDPBD158304-5GEA 19 EUR
BLDPBD158304-5G BLDPBD158304-100G BLDPBD158304-1G BLDPBD158304-25G BLDPBD158304-10G BLDPBD158304-500G
tert-Butyl-2,4-dioxopiperidine-1-carboxylate 97%
tert-Butyl-2,4-dioxopiperidine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₅NO₄
Molecular Weight: 213,2304 g/mol
Lagertemperatur: Ambient
MDL: MFCD10566071
CAS-Nummer: 845267-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR