1-(6-Methoxy-2-naphthyl)ethanon 98%

Synonyme: 6'-Methoxy-2'-acetonaphthone

BD1583-10G BD1583-25G BD1583-500G BD1583-100G
BLDPBD1583-10GEA 7 EUR
BLDPBD1583-10G BLDPBD1583-25G BLDPBD1583-500G BLDPBD1583-100G
1-(6-Methoxy-2-naphthyl)ethanon 98%
2-Acetyl-6-methoxynaphthalene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₂O₂
Molecular Weight: 200,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD00021643
CAS-Nummer: 3900-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR