tert-Butyl 7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate 95%

BD158295-250MG BD158295-1G BD158295-5G
BLDPBD158295-250MGEA 10 EUR
BLDPBD158295-250MG BLDPBD158295-1G BLDPBD158295-5G
tert-Butyl 7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate 95%
tert-Butyl 7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₇N₃O₂
Molecular Weight: 235,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD12407806
CAS-Nummer: 192869-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR